missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 608 Organochlorine Pesticide Mixture 391 20 μg/mL in Isooctane, Dr. Ehrenstorfer

Product Code. 30312028
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30312028 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312028 Supplier Dr. Ehrenstorfer Supplier No. DREA50000391IO
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣delta-HCH∣Dieldrin∣Endosulfan-sulfate∣Endrin∣Endrin-aldehyde∣gamma-HCH∣Heptachlor∣Heptachlor-exo-epoxide (cis-, isomer B)
CAS 72-54-8, 72-55-9, 50-29-3, 309-00-2, 959-98-8, 319-84-6
Analyte or Component Names 4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣delta-HCH∣Dieldrin∣Endosulfan-sulfate∣Endrin∣Endrin-aldehyde∣gamma-HCH∣Heptachlor∣Heptachlor-exo-epoxide (cis-, isomer B)
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Isooctane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 4,4'-DDD 20 μg/mL∣4,4'-DDE 20 μg/mL∣4,4'-DDT 20 μg/mL∣Aldrin 20 μg/mL∣alpha-Endosulfan 20 μg/mL∣alpha-HCH 20 μg/mL∣beta-Endosulfan 20 μg/mL∣beta-HCH 20 μg/mL∣delta-HCH 20 μg/mL∣Dieldrin 20 μg/mL∣Endosulfan-sulfate 20 μg/mL∣Endrin 20 μg/mL∣Endrin-aldehyde 20 μg/mL∣gamma-HCH 20 μg/mL∣Heptachlor 20 μg/mL∣Heptachlor-exo-epoxide (cis-, isomer B) 20 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.