missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 625.1 Surrogate Mixture 1000 μg/mL in Methanol, Dr. Ehrenstorfer
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
Specifications
Specifications
| Chemical Name or Material | 2-Fluorophenol∣2-Nitrophenol-d4∣Aniline D5∣Nitrobenzene D5∣Phenol-d5∣Pyridine D5 |
| CAS | 367-12-4, 93951-78-1, 4165-61-1, 4165-60-0, 4165-62-2, 7291-22-7 |
| Analyte or Component Names | 2-Fluorophenol∣2-Nitrophenol-d4∣Aniline D5∣Nitrobenzene D5∣Phenol-d5∣Pyridine D5 |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Methanol |
| Quantity | 5 x 1 mL |
| Concentration or Composition (by Analyte or Components) | 2-Fluorophenol 1000 μg/ml∣2-Nitrophenol-d4 1000 μg/ml∣Aniline D5 1000 μg/ml∣Nitrobenzene D5 1000 μg/ml∣Phenol-d5 1000 μg/ml∣Pyridine D5 1000 μg/ml |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction