missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 625 SV Calibration Mixture 660 1000 μg/mL in Benzene:Dichloromethane, Dr. Ehrenstorfer

Product Code. 30314836
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30314836 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30314836 Supplier Dr. Ehrenstorfer Supplier No. DREA50000660BD
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2,4-Trichlorobenzene∣1,2-Dichlorobenzene∣1,3-Dichlorobenzene∣1,4-Dichlorobenzene∣2,4,6-Trichlorophenol∣2,4-Dichlorophenol∣2,4-Dimethylphenol∣2,4-Dinitrophenol∣2,4-Dinitrotoluene∣2,6-Dinitrotoluene∣2-Chloronaphthalene∣2-Chlorophenol∣2-Methyl-4,6-dinitrophenol∣2-Nitrophenol∣4-Chloro-3-methylphenol∣4-Chlorophenyl phenyl ether∣4-Nitrophenol∣Acenaphthene∣Acenaphthylene∣Anthracene∣Azobenzene∣Benz[a]anthracene∣Benzo(k)fluoranthene∣Benzo[a]pyrene∣Benzo[b]fluoranthene∣Benzo[ghi]perylene∣Bis-(2-chloro-1-methylethyl)ether∣Bis(2-chloroethoxy)methane∣Bis-(2-chloroethyl)ether∣Carbazole∣Chrysene∣Dibenzo(a,h)anthracene∣Dibutyl phthalate∣Diethyl phthalate∣Di-n-octyl phthalate∣Fluoranthene∣Fluorene∣Hexachlorobenzene∣Hexachlorobutadiene∣Hexachlorocyclopentadiene∣Hexachloroethane∣Indeno[1,2,3-cd]pyrene∣Isophorone∣Naphthalene∣Nitrobenzene∣N-Nitrosodimethylamine∣N-Nitroso-di-n-propylamine∣PBDE 3 (4-Bromodiphenyl Ether)∣Pentachlorophenol∣Phenanthrene∣Phenol∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-methyl ester∣Pyrene
CAS 120-82-1, 95-50-1, 541-73-1, 106-46-7, 88-06-2, 120-83-2
Analyte or Component Names 1,2,4-Trichlorobenzene∣1,2-Dichlorobenzene∣1,3-Dichlorobenzene∣1,4-Dichlorobenzene∣2,4,6-Trichlorophenol∣2,4-Dichlorophenol∣2,4-Dimethylphenol∣2,4-Dinitrophenol∣2,4-Dinitrotoluene∣2,6-Dinitrotoluene∣2-Chloronaphthalene∣2-Chlorophenol∣2-Methyl-4,6-dinitrophenol∣2-Nitrophenol∣4-Chloro-3-methylphenol∣4-Chlorophenyl phenyl ether∣4-Nitrophenol∣Acenaphthene∣Acenaphthylene∣Anthracene∣Azobenzene∣Benz[a]anthracene∣Benzo(k)fluoranthene∣Benzo[a]pyrene∣Benzo[b]fluoranthene∣Benzo[ghi]perylene∣Bis-(2-chloro-1-methylethyl)ether∣Bis(2-chloroethoxy)methane∣Bis-(2-chloroethyl)ether∣Carbazole∣Chrysene∣Dibenzo(a,h)anthracene∣Dibutyl phthalate∣Diethyl phthalate∣Di-n-octyl phthalate∣Fluoranthene∣Fluorene∣Hexachlorobenzene∣Hexachlorobutadiene∣Hexachlorocyclopentadiene∣Hexachloroethane∣Indeno[1,2,3-cd]pyrene∣Isophorone∣Naphthalene∣Nitrobenzene∣N-Nitrosodimethylamine∣N-Nitroso-di-n-propylamine∣PBDE 3 (4-Bromodiphenyl Ether)∣Pentachlorophenol∣Phenanthrene∣Phenol∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-methyl ester∣Pyrene
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Benzene/Dichloromethane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1,2,4-Trichlorobenzene 1000 μg/mL∣1,2-Dichlorobenzene 1000 μg/mL∣1,3-Dichlorobenzene 1000 μg/mL∣1,4-Dichlorobenzene 1000 μg/mL∣2,4,6-Trichlorophenol 1000 μg/mL∣2,4-Dichlorophenol 1000 μg/mL∣2,4-Dimethylphenol 1000 μg/mL∣2,4-Dinitrophenol 1000 μg/mL∣2,4-Dinitrotoluene 1000 μg/mL∣2,6-Dinitrotoluene 1000 μg/mL∣2-Chloronaphthalene 1000 μg/mL∣2-Chlorophenol 1000 μg/mL∣2-Methyl-4,6-dinitrophenol 1000 μg/mL∣2-Nitrophenol 1000 μg/mL∣4-Chloro-3-methylphenol 1000 μg/mL∣4-Chlorophenyl phenyl ether 1000 μg/mL∣4-Nitrophenol 1000 μg/mL∣Acenaphthene 1000 μg/mL∣Acenaphthylene 1000 μg/mL∣Anthracene 1000 μg/mL∣Azobenzene 1000 μg/mL∣Benz[a]anthracene 1000 μg/mL∣Benzo(k)fluoranthene 1000 μg/mL∣Benzo[a]pyrene 1000 μg/mL∣Benzo[b]fluoranthene 1000 μg/mL∣Benzo[ghi]perylene 1000 μg/mL∣Bis-(2-chloro-1-methylethyl)ether 1000 μg/mL∣Bis(2-chloroethoxy)methane 1000 μg/mL∣Bis-(2-chloroethyl)ether 1000 μg/mL∣Carbazole 1000 μg/mL∣Chrysene 1000 μg/mL∣Dibenzo(a,h)anthracene 1000 μg/mL∣Dibutyl phthalate 1000 μg/mL∣Diethyl phthalate 1000 μg/mL∣Di-n-octyl phthalate 1000 μg/mL∣Fluoranthene 1000 μg/mL∣Fluorene 1000 μg/mL∣Hexachlorobenzene 1000 μg/mL∣Hexachlorobutadiene 1000 μg/mL∣Hexachlorocyclopentadiene 1000 μg/mL∣Hexachloroethane 1000 μg/mL∣Indeno[1,2,3-cd]pyrene 1000 μg/mL∣Isophorone 1000 μg/mL∣Naphthalene 1000 μg/mL∣Nitrobenzene 1000 μg/mL∣N-Nitrosodimethylamine 1000 μg/mL∣N-Nitroso-di-n-propylamine 1000 μg/mL∣PBDE 3 (4-Bromodiphenyl Ether) 1000 μg/mL∣Pentachlorophenol 1000 μg/mL∣Phenanthrene 1000 μg/mL∣Phenol 1000 μg/mL∣Phthalic acid, benzylbutyl ester 1000 μg/mL∣Phthalic acid, bis-2-ethylhexyl ester 1000 μg/mL∣Phthalic acid, bis-methyl ester 1000 μg/mL∣Pyrene 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.