missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 8020 Aromatic VOC Mixture 416 200 μg/mL in Methanol, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | 1,2-Dichlorobenzene∣1,3-Dichlorobenzene∣1,4-Dichlorobenzene∣Benzene∣Chlorobenzene∣Ethylbenzene∣m-Xylene (1,3-Dimethylbenzene)∣o-Xylene (1,2-Dimethylbenzene)∣p-Xylene (1,4-Dimethylbenzene)∣Toluene |
| CAS | 95-50-1, 541-73-1, 106-46-7, 71-43-2, 108-90-7, 100-41-4 |
| Analyte or Component Names | 1,2-Dichlorobenzene∣1,3-Dichlorobenzene∣1,4-Dichlorobenzene∣Benzene∣Chlorobenzene∣Ethylbenzene∣m-Xylene (1,3-Dimethylbenzene)∣o-Xylene (1,2-Dimethylbenzene)∣p-Xylene (1,4-Dimethylbenzene)∣Toluene |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Methanol |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 1,2-Dichlorobenzene 200 μg/mL∣1,3-Dichlorobenzene 200 μg/mL∣1,4-Dichlorobenzene 200 μg/mL∣Benzene 200 μg/mL∣Chlorobenzene 200 μg/mL∣Ethylbenzene 200 μg/mL∣m-Xylene (1,3-Dimethylbenzene) 200 μg/mL∣o-Xylene (1,2-Dimethylbenzene) 200 μg/mL∣p-Xylene (1,4-Dimethylbenzene) 200 μg/mL∣Toluene 200 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction