missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 8061 Phthalate Mixture 438 1000 μg/mL in n-Hexane, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Dibutyl phthalate∣Diethyl phthalate∣Di-n-octyl phthalate∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethoxyethyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-4-methyl-2-pentyl ester∣Phthalic acid, bis-cyclohexyl ester∣Phthalic acid, bis-hexyl ester∣Phthalic acid, bis-iso-butyl ester∣Phthalic acid, bis-methyl ester∣Phthalic acid, bis-methylglycol ester∣Phthalic acid, bis-nonyl ester∣Phthalic acid, bis-n-pentyl ester∣Phthalic acid,bis-butoxyethyl ester |
| CAS | 84-74-2, 84-66-2, 117-84-0, 85-68-7, 605-54-9, 117-81-7 |
| Analyte or Component Names | Dibutyl phthalate∣Diethyl phthalate∣Di-n-octyl phthalate∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethoxyethyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-4-methyl-2-pentyl ester∣Phthalic acid, bis-cyclohexyl ester∣Phthalic acid, bis-hexyl ester∣Phthalic acid, bis-iso-butyl ester∣Phthalic acid, bis-methyl ester∣Phthalic acid, bis-methylglycol ester∣Phthalic acid, bis-nonyl ester∣Phthalic acid, bis-n-pentyl ester∣Phthalic acid,bis-butoxyethyl ester |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Hexane |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Dibutyl phthalate 1000 μg/mL∣Diethyl phthalate 1000 μg/mL∣Di-n-octyl phthalate 1000 μg/mL∣Phthalic acid, benzylbutyl ester 1000 μg/mL∣Phthalic acid, bis-2-ethoxyethyl ester 1000 μg/mL∣Phthalic acid, bis-2-ethylhexyl ester 1000 μg/mL∣Phthalic acid, bis-4-methyl-2-pentyl ester 1000 μg/mL∣Phthalic acid, bis-cyclohexyl ester 1000 μg/mL∣Phthalic acid, bis-hexyl ester 1000 μg/mL∣Phthalic acid, bis-iso-butyl ester 1000 μg/mL∣Phthalic acid, bis-methyl ester 1000 μg/mL∣Phthalic acid, bis-methylglycol ester 1000 μg/mL∣Phthalic acid, bis-nonyl ester 1000 μg/mL∣Phthalic acid, bis-n-pentyl ester 1000 μg/mL∣Phthalic acid,bis-butoxyethyl ester 1000 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?