missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 8080A/8081 Organochlorine Pesticide Mixture 616 200 μg/mL in Hexane:Toluene, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 2,4'-DDT∣4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-HCH∣beta-HCH∣delta-HCH∣Dieldrin∣Endrin∣gamma-HCH∣Heptachlor∣Heptachlor-exo-epoxide (cis-, isomer B)∣Hexachlorobenzene |
| CAS | 789-02-6, 72-54-8, 72-55-9, 50-29-3, 309-00-2, 319-84-6 |
| Analyte or Component Names | 2,4'-DDT∣4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-HCH∣beta-HCH∣delta-HCH∣Dieldrin∣Endrin∣gamma-HCH∣Heptachlor∣Heptachlor-exo-epoxide (cis-, isomer B)∣Hexachlorobenzene |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Hexane/Toluene |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 2,4'-DDT 200 μg/mL∣4,4'-DDD 200 μg/mL∣4,4'-DDE 200 μg/mL∣4,4'-DDT 200 μg/mL∣Aldrin 200 μg/mL∣alpha-HCH 200 μg/mL∣beta-HCH 200 μg/mL∣delta-HCH 200 μg/mL∣Dieldrin 200 μg/mL∣Endrin 200 μg/mL∣gamma-HCH 200 μg/mL∣Heptachlor 200 μg/mL∣Heptachlor-exo-epoxide (cis-, isomer B) 200 μg/mL∣Hexachlorobenzene 200 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?