missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 8080A Organochlorine Pesticide Mixture 613 1000 μg/mL in Hexane:Toluene, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 2,4'-DDD∣2,4'-DDE∣2,4'-DDT |
| CAS | 53-19-0, 3424-82-6, 789-02-6 |
| Analyte or Component Names | 2,4'-DDD∣2,4'-DDE∣2,4'-DDT |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Hexane/Toluene |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 2,4'-DDD 1000 μg/mL∣2,4'-DDE 1000 μg/mL∣2,4'-DDT 1000 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?