missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 8091 Non-RCRA Analyte Mixture 426 1000 μg/mL in Isooctane:Toluene, Dr. Ehrenstorfer

Product Code. 30320152
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30320152 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30320152 Supplier Dr. Ehrenstorfer Supplier No. DREA50000426IT
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3,5-Trichloro-2-nitrobenzene∣1-Chloro-2,4-dinitrobenzene∣1-Chloro-2-nitrobenzene∣1-Chloro-3,4-dinitrobenzene∣1-Chloro-4-nitrobenzene∣2,3,4,5-Tetrachloronitrobenzene∣2,3,4-Trichloronitrobenzene∣2,3,5,6-Tetrachloronitrobenzene∣2,3-Dichloronitrobenzene∣2,4,5-Trichloronitrobenzene∣2,4-Dichloronitrobenzene∣2,5-Dichloronitrobenzene∣2-Chloro-6-nitrotoluene∣3,4-Dichloronitrobenzene∣3,5-Dichloronitrobenzene∣4-Chloro-2-nitrotoluene∣4-Chloro-3-nitrotoluene
CAS 18708-70-8, 97-00-7, 88-73-3, 610-40-2, 100-00-5, 879-39-0
Analyte or Component Names 1,3,5-Trichloro-2-nitrobenzene∣1-Chloro-2,4-dinitrobenzene∣1-Chloro-2-nitrobenzene∣1-Chloro-3,4-dinitrobenzene∣1-Chloro-4-nitrobenzene∣2,3,4,5-Tetrachloronitrobenzene∣2,3,4-Trichloronitrobenzene∣2,3,5,6-Tetrachloronitrobenzene∣2,3-Dichloronitrobenzene∣2,4,5-Trichloronitrobenzene∣2,4-Dichloronitrobenzene∣2,5-Dichloronitrobenzene∣2-Chloro-6-nitrotoluene∣3,4-Dichloronitrobenzene∣3,5-Dichloronitrobenzene∣4-Chloro-2-nitrotoluene∣4-Chloro-3-nitrotoluene
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Isooctane/Toluene
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1,3,5-Trichloro-2-nitrobenzene 1000 μg/mL∣1-Chloro-2,4-dinitrobenzene 1000 μg/mL∣1-Chloro-2-nitrobenzene 1000 μg/mL∣1-Chloro-3,4-dinitrobenzene 1000 μg/mL∣1-Chloro-4-nitrobenzene 1000 μg/mL∣2,3,4,5-Tetrachloronitrobenzene 1000 μg/mL∣2,3,4-Trichloronitrobenzene 1000 μg/mL∣2,3,5,6-Tetrachloronitrobenzene 1000 μg/mL∣2,3-Dichloronitrobenzene 1000 μg/mL∣2,4,5-Trichloronitrobenzene 1000 μg/mL∣2,4-Dichloronitrobenzene 1000 μg/mL∣2,5-Dichloronitrobenzene 1000 μg/mL∣2-Chloro-6-nitrotoluene 1000 μg/mL∣3,4-Dichloronitrobenzene 1000 μg/mL∣3,5-Dichloronitrobenzene 1000 μg/mL∣4-Chloro-2-nitrotoluene 1000 μg/mL∣4-Chloro-3-nitrotoluene 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.