missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 8141 Organophosphorus Pesticide Mixture 428 200 μg/mL in n-Hexane, Dr. Ehrenstorfer

Product Code. 30324136
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30324136 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324136 Supplier Dr. Ehrenstorfer Supplier No. DREA50000428HE
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Azinphos-ethyl∣Carbophenothion∣Chlorfenvinphos∣Dioxathion∣Ethion∣Famphur∣Leptophos∣Phosmet∣Phosphamidon∣Terbufos
CAS 2642-71-9, 786-19-6, 470-90-6, 78-34-2, 563-12-2, 52-85-7
Analyte or Component Names Azinphos-ethyl∣Carbophenothion∣Chlorfenvinphos∣Dioxathion∣Ethion∣Famphur∣Leptophos∣Phosmet∣Phosphamidon∣Terbufos
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix n-Hexane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Azinphos-ethyl 200 μg/mL∣Carbophenothion 200 μg/mL∣Chlorfenvinphos 200 μg/mL∣Dioxathion 200 μg/mL∣Ethion 200 μg/mL∣Famphur 200 μg/mL∣Leptophos 200 μg/mL∣Phosmet 200 μg/mL∣Phosphamidon 200 μg/mL∣Terbufos 200 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.