missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 8260 Acetate Mixture 2000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30310346
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30310346 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30310346 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000415ME
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetic acid-isopropyl ester∣Amyl Acetate∣Butyl Acetate∣Ethyl acetate∣Methyl Acetate∣Propyl Acetate∣Vinylacetate
CAS 108-21-4, 628-63-7, 123-86-4, 141-78-6, 79-20-9, 109-60-4
Analyte or Component Names Acetic acid-isopropyl ester∣Amyl Acetate∣Butyl Acetate∣Ethyl acetate∣Methyl Acetate∣Propyl Acetate∣Vinylacetate
Shipping Condition Ice pack (-20°C)
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Acetic acid-isopropyl ester 2000 μg/mL∣Amyl Acetate 2000 μg/mL∣Butyl Acetate 2000 μg/mL∣Ethyl acetate 2000 μg/mL∣Methyl Acetate 2000 μg/mL∣Propyl Acetate 2000 μg/mL∣Vinylacetate 2000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.