missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 8260 Matrix Spike Mixture 2500 μg/mL in Methanol, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 1,1-Dichloroethene∣Benzene∣Chlorobenzene∣Toluene∣Trichloroethene |
| CAS | 75-35-4, 71-43-2, 108-90-7, 108-88-3, 79-01-6 |
| Analyte or Component Names | 1,1-Dichloroethene∣Benzene∣Chlorobenzene∣Toluene∣Trichloroethene |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Methanol |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 1,1-Dichloroethene 2500 μg/mL∣Benzene 2500 μg/mL∣Chlorobenzene 2500 μg/mL∣Toluene 2500 μg/mL∣Trichloroethene 2500 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?