missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 8260 VOC Mixture 565 200 μg/mL in Methanol, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | 1-Chlorohexane (Hexylchloride)∣2-Butanone∣2-Chloroethyl-vinylether∣2-Hexanone∣4-Methyl-2-pentanone (MIBK)∣Acetone∣Acrylonitrile∣alpha-Methylstyrene∣Carbon disulfide∣Cyclohexane∣Dimethyl sulfide∣Isoprene∣Methacrylic acid-ethyl ester∣Methyl Acetate∣Methyl Disulfide∣Methyl iodide∣Methyl tert-butyl ether∣Methylcyclohexane∣n-Hexane∣trans-1,4-Dichloro-2-butene |
| CAS | 544-10-5, 78-93-3, 110-75-8, 591-78-6, 108-10-1, 67-64-1 |
| Analyte or Component Names | 1-Chlorohexane (Hexylchloride)∣2-Butanone∣2-Chloroethyl-vinylether∣2-Hexanone∣4-Methyl-2-pentanone (MIBK)∣Acetone∣Acrylonitrile∣alpha-Methylstyrene∣Carbon disulfide∣Cyclohexane∣Dimethyl sulfide∣Isoprene∣Methacrylic acid-ethyl ester∣Methyl Acetate∣Methyl Disulfide∣Methyl iodide∣Methyl tert-butyl ether∣Methylcyclohexane∣n-Hexane∣trans-1,4-Dichloro-2-butene |
| Shipping Condition | -18°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Methanol |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 1-Chlorohexane (Hexylchloride) 200 μg/mL∣2-Butanone 200 μg/mL∣2-Chloroethyl-vinylether 200 μg/mL∣2-Hexanone 200 μg/mL∣4-Methyl-2-pentanone (MIBK) 200 μg/mL∣Acetone 200 μg/mL∣Acrylonitrile 200 μg/mL∣alpha-Methylstyrene 200 μg/mL∣Carbon disulfide 200 μg/mL∣Cyclohexane 200 μg/mL∣Dimethyl sulfide 200 μg/mL∣Isoprene 200 μg/mL∣Methacrylic acid-ethyl ester 200 μg/mL∣Methyl Acetate 200 μg/mL∣Methyl Disulfide 200 μg/mL∣Methyl iodide 200 μg/mL∣Methyl tert-butyl ether 200 μg/mL∣Methylcyclohexane 200 μg/mL∣n-Hexane 200 μg/mL∣trans-1,4-Dichloro-2-butene 200 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction