missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 8270 Appendix IX Mixture 1 Minus Phentermine 2000 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30312910
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30312910 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312910 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000444DI

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,4-Phenylenediamine (1,4-Diaminobenzene)∣1-Aminonaphthalene∣2-Acetamidofluoren∣2-Amino-4-nitrotoluene (2-Methyl-5-nitroaniline)∣2-Aminonaphthalene∣2-Picolin (2-Methylpyridine)∣4-Aminobiphenyl∣4-Nitrosomorpholine∣Isosafrole∣N-Nitroso-diethylamine∣N-Nitroso-di-n-butylamine∣N-Nitrosomethylethylamine∣N-Nitrosopiperidine∣N-Nitrosopyrrolidine∣o-Toluidine∣Sudan Yellow
CAS 106-50-3, 134-32-7, 53-96-3, 99-55-8, 91-59-8, 109-06-8
Analyte or Component Names 1,4-Phenylenediamine (1,4-Diaminobenzene)∣1-Aminonaphthalene∣2-Acetamidofluoren∣2-Amino-4-nitrotoluene (2-Methyl-5-nitroaniline)∣2-Aminonaphthalene∣2-Picolin (2-Methylpyridine)∣4-Aminobiphenyl∣4-Nitrosomorpholine∣Isosafrole∣N-Nitroso-diethylamine∣N-Nitroso-di-n-butylamine∣N-Nitrosomethylethylamine∣N-Nitrosopiperidine∣N-Nitrosopyrrolidine∣o-Toluidine∣Sudan Yellow
Shipping Condition Ice pack (-20°C)
Certifications/Compliance ISO 17034
Solvent or Matrix Dichloromethane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1,4-Phenylenediamine (1,4-Diaminobenzene) 2000 μg/mL∣1-Aminonaphthalene 2000 μg/mL∣2-Acetamidofluoren 2000 μg/mL∣2-Amino-4-nitrotoluene (2-Methyl-5-nitroaniline) 2000 μg/mL∣2-Aminonaphthalene 2000 μg/mL∣2-Picolin (2-Methylpyridine) 2000 μg/mL∣4-Aminobiphenyl 2000 μg/mL∣4-Nitrosomorpholine 2000 μg/mL∣Isosafrole 2000 μg/mL∣N-Nitroso-diethylamine 2000 μg/mL∣N-Nitroso-di-n-butylamine 2000 μg/mL∣N-Nitrosomethylethylamine 2000 μg/mL∣N-Nitrosopiperidine 2000 μg/mL∣N-Nitrosopyrrolidine 2000 μg/mL∣o-Toluidine 2000 μg/mL∣Sudan Yellow 2000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.