missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 8270 Appendix IX Mixture 1 with Phentermine 2000 μg/mL in Dichloromethane, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | 1,4-Phenylenediamine (1,4-Diaminobenzene)∣1-Aminonaphthalene∣2-Acetamidofluoren∣2-Amino-4-nitrotoluene (2-Methyl-5-nitroaniline)∣2-Aminonaphthalene∣2-Picolin (2-Methylpyridine)∣4-Aminobiphenyl∣4-Nitrosomorpholine∣Isosafrole∣N-Nitroso-diethylamine∣N-Nitroso-di-n-butylamine∣N-Nitrosomethylethylamine∣N-Nitrosopiperidine∣N-Nitrosopyrrolidine∣o-Toluidine∣Phentermine∣Sudan Yellow |
| CAS | 106-50-3, 134-32-7, 53-96-3, 99-55-8, 91-59-8, 109-06-8 |
| Analyte or Component Names | 1,4-Phenylenediamine (1,4-Diaminobenzene)∣1-Aminonaphthalene∣2-Acetamidofluoren∣2-Amino-4-nitrotoluene (2-Methyl-5-nitroaniline)∣2-Aminonaphthalene∣2-Picolin (2-Methylpyridine)∣4-Aminobiphenyl∣4-Nitrosomorpholine∣Isosafrole∣N-Nitroso-diethylamine∣N-Nitroso-di-n-butylamine∣N-Nitrosomethylethylamine∣N-Nitrosopiperidine∣N-Nitrosopyrrolidine∣o-Toluidine∣Phentermine∣Sudan Yellow |
| Shipping Condition | Ice pack (-20°C) |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dichloromethane |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 1,4-Phenylenediamine (1,4-Diaminobenzene) 2000 μg/mL∣1-Aminonaphthalene 2000 μg/mL∣2-Acetamidofluoren 2000 μg/mL∣2-Amino-4-nitrotoluene (2-Methyl-5-nitroaniline) 2000 μg/mL∣2-Aminonaphthalene 2000 μg/mL∣2-Picolin (2-Methylpyridine) 2000 μg/mL∣4-Aminobiphenyl 2000 μg/mL∣4-Nitrosomorpholine 2000 μg/mL∣Isosafrole 2000 μg/mL∣N-Nitroso-diethylamine 2000 μg/mL∣N-Nitroso-di-n-butylamine 2000 μg/mL∣N-Nitrosomethylethylamine 2000 μg/mL∣N-Nitrosopiperidine 2000 μg/mL∣N-Nitrosopyrrolidine 2000 μg/mL∣o-Toluidine 2000 μg/mL∣Phentermine 2000 μg/mL∣Sudan Yellow 2000 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction