missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 8270 B/N Mixture 2 1000 μg/ml in Benz:MeCl2:ACCN 4:4:2, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 1,2,4-Trichlorobenzene∣1,2-Dichlorobenzene∣1,3-Dichlorobenzene∣1,4-Dichlorobenzene∣2,4-Dinitrotoluene∣2,6-Dinitrotoluene∣2-Chloronaphthalene∣4-Chlorophenyl phenyl ether∣Acenaphthene∣Acenaphthylene∣Anthracene∣Azobenzene∣Benz[a]anthracene∣Benzo(k)fluoranthene∣Benzo[a]pyrene∣Benzo[b]fluoranthene∣Benzo[ghi]perylene∣Bis-(2-chloro-1-methylethyl)ether∣Bis(2-chloroethoxy)methane∣Bis-(2-chloroethyl)ether∣Chrysene∣Dibenzo(a,h)anthracene∣Dibutyl phthalate∣Diethyl phthalate∣Di-n-octyl phthalate∣Fluoranthene∣Fluorene∣Hexachlorobenzene∣Hexachlorobutadiene∣Hexachlorocyclopentadiene∣Hexachloroethane∣Indeno[1,2,3-cd]pyrene∣Isophorone∣Naphthalene∣Nitrobenzene∣N-Nitrosodimethylamine∣N-Nitroso-di-n-propylamine∣N-Nitroso-diphenylamine∣PBDE 3 (4-Bromodiphenyl Ether)∣Phenanthrene∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-methyl ester∣Pyrene |
| CAS | 120-82-1, 95-50-1, 541-73-1, 106-46-7, 121-14-2, 606-20-2 |
| Analyte or Component Names | 1,2,4-Trichlorobenzene∣1,2-Dichlorobenzene∣1,3-Dichlorobenzene∣1,4-Dichlorobenzene∣2,4-Dinitrotoluene∣2,6-Dinitrotoluene∣2-Chloronaphthalene∣4-Chlorophenyl phenyl ether∣Acenaphthene∣Acenaphthylene∣Anthracene∣Azobenzene∣Benz[a]anthracene∣Benzo(k)fluoranthene∣Benzo[a]pyrene∣Benzo[b]fluoranthene∣Benzo[ghi]perylene∣Bis-(2-chloro-1-methylethyl)ether∣Bis(2-chloroethoxy)methane∣Bis-(2-chloroethyl)ether∣Chrysene∣Dibenzo(a,h)anthracene∣Dibutyl phthalate∣Diethyl phthalate∣Di-n-octyl phthalate∣Fluoranthene∣Fluorene∣Hexachlorobenzene∣Hexachlorobutadiene∣Hexachlorocyclopentadiene∣Hexachloroethane∣Indeno[1,2,3-cd]pyrene∣Isophorone∣Naphthalene∣Nitrobenzene∣N-Nitrosodimethylamine∣N-Nitroso-di-n-propylamine∣N-Nitroso-diphenylamine∣PBDE 3 (4-Bromodiphenyl Ether)∣Phenanthrene∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-methyl ester∣Pyrene |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Benzene/Dichloromethane/Acetonitrile |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 1,2,4-Trichlorobenzene 1000 μg/mL∣1,2-Dichlorobenzene 1000 μg/mL∣1,3-Dichlorobenzene 1000 μg/mL∣1,4-Dichlorobenzene 1000 μg/mL∣2,4-Dinitrotoluene 1000 μg/mL∣2,6-Dinitrotoluene 1000 μg/mL∣2-Chloronaphthalene 1000 μg/mL∣4-Chlorophenyl phenyl ether 1000 μg/mL∣Acenaphthene 1000 μg/mL∣Acenaphthylene 1000 μg/mL∣Anthracene 1000 μg/mL∣Azobenzene 1000 μg/mL∣Benz[a]anthracene 1000 μg/mL∣Benzo(k)fluoranthene 1000 μg/mL∣Benzo[a]pyrene 1000 μg/mL∣Benzo[b]fluoranthene 1000 μg/mL∣Benzo[ghi]perylene 1000 μg/mL∣Bis-(2-chloro-1-methylethyl)ether 1000 μg/mL∣Bis(2-chloroethoxy)methane 1000 μg/mL∣Bis-(2-chloroethyl)ether 1000 μg/mL∣Chrysene 1000 μg/mL∣Dibenzo(a,h)anthracene 1000 μg/mL∣Dibutyl phthalate 1000 μg/mL∣Diethyl phthalate 1000 μg/mL∣Di-n-octyl phthalate 1000 μg/mL∣Fluoranthene 1000 μg/mL∣Fluorene 1000 μg/mL∣Hexachlorobenzene 1000 μg/mL∣Hexachlorobutadiene 1000 μg/mL∣Hexachlorocyclopentadiene 1000 μg/mL∣Hexachloroethane 1000 μg/mL∣Indeno[1,2,3-cd]pyrene 1000 μg/mL∣Isophorone 1000 μg/mL∣Naphthalene 1000 μg/mL∣Nitrobenzene 1000 μg/mL∣N-Nitrosodimethylamine 1000 μg/mL∣N-Nitroso-di-n-propylamine 1000 μg/mL∣N-Nitroso-diphenylamine 1000 μg/mL∣PBDE 3 (4-Bromodiphenyl Ether) 1000 μg/mL∣Phenanthrene 1000 μg/mL∣Phthalic acid, benzylbutyl ester 1000 μg/mL∣Phthalic acid, bis-2-ethylhexyl ester 1000 μg/mL∣Phthalic acid, bis-methyl ester 1000 μg/mL∣Pyrene 1000 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?