missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 8270 BNA Surrogate Mixture 5000 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30326091
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30326091 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326091 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000432DI
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4,6-Tribromophenol∣2-Fluorobiphenyl∣2-Fluorophenol∣Nitrobenzene D5∣Phenol-d5∣p-Terphenyl D14∣Pyrene-d10
CAS 118-79-6, 321-60-8, 367-12-4, 4165-60-0, 4165-62-2, 1718-51-0
Analyte or Component Names 2,4,6-Tribromophenol∣2-Fluorobiphenyl∣2-Fluorophenol∣Nitrobenzene D5∣Phenol-d5∣p-Terphenyl D14∣Pyrene-d10
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dichloromethane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 2,4,6-Tribromophenol 5000 μg/mL∣2-Fluorobiphenyl 5000 μg/mL∣2-Fluorophenol 5000 μg/mL∣Nitrobenzene D5 5000 μg/mL∣Phenol-d5 5000 μg/mL∣p-Terphenyl D14 5000 μg/mL∣Pyrene-d10 5000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.