missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 8270 Calibration Mixture 500-1000 μg/mL in Dichloromethane, Dr. Ehrenstorfer
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
Specifications
Specifications
| Chemical Name or Material | 1,2,4-Trichlorobenzene∣1,2-Dichlorobenzene∣1,2-Dinitrobenzene∣1,3-Dichlorobenzene∣1,3-Dinitrobenzene∣1,4-Dichlorobenzene∣1,4-Dinitrobenzene∣1-Methylnaphthalene∣2,3,4,6-Tetrachlorophenol∣2,3,5,6-Tetrachlorophenol∣2,4,5-Trichlorophenol∣2,4,6-Trichlorophenol∣2,4-Dichlorophenol∣2,4-Dimethylphenol∣2,4-Dinitrophenol∣2,4-Dinitrotoluene∣2,6-Dinitrotoluene∣2-Chloronaphthalene∣2-Chlorophenol∣2-Methyl-4,6-dinitrophenol∣2-Methylnaphthalene∣2-Methylphenol∣2-Nitroaniline∣2-Nitrophenol∣3-Methylphenol (m-Cresol)∣3-Nitroaniline∣4-Chloro-3-methylphenol∣4-Chloroaniline∣4-Chlorophenyl phenyl ether∣4-Methylphenol (p-Cresol)∣4-Nitroaniline∣4-Nitrophenol∣Acenaphthene∣Acenaphthylene∣Adipic Acid Bis(2-ethylhexyl) Ester∣Aniline∣Anthracene∣Azobenzene∣Benz[a]anthracene∣Benzo(k)fluoranthene∣Benzo[a]pyrene∣Benzo[b]fluoranthene∣Benzo[ghi]perylene∣Benzyl alcohol∣Bis-(2-chloro-1-methylethyl)ether∣Bis(2-chloroethoxy)methane∣Bis-(2-chloroethyl)ether∣Carbazole∣Chrysene∣Dibenzo(a,h)anthracene∣Dibenzofuran∣Dibutyl phthalate∣Diethyl phthalate∣Di-n-octyl phthalate∣Diphenylamine∣Fluoranthene∣Fluorene∣Hexachlorobenzene∣Hexachlorobutadiene∣Hexachlorocyclopentadiene∣Hexachloroethane∣Indeno[1,2,3-cd]pyrene∣Isophorone∣Naphthalene∣Nitrobenzene∣N-Nitrosodimethylamine∣N-Nitroso-di-n-propylamine∣PBDE 3 (4-Bromodiphenyl Ether)∣Pentachlorophenol∣Phenanthrene∣Phenol∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-methyl ester∣Pyrene∣Pyridine |
| CAS | 120-82-1, 95-50-1, 528-29-0, 541-73-1, 99-65-0, 106-46-7 |
| Analyte or Component Names | 1,2,4-Trichlorobenzene∣1,2-Dichlorobenzene∣1,2-Dinitrobenzene∣1,3-Dichlorobenzene∣1,3-Dinitrobenzene∣1,4-Dichlorobenzene∣1,4-Dinitrobenzene∣1-Methylnaphthalene∣2,3,4,6-Tetrachlorophenol∣2,3,5,6-Tetrachlorophenol∣2,4,5-Trichlorophenol∣2,4,6-Trichlorophenol∣2,4-Dichlorophenol∣2,4-Dimethylphenol∣2,4-Dinitrophenol∣2,4-Dinitrotoluene∣2,6-Dinitrotoluene∣2-Chloronaphthalene∣2-Chlorophenol∣2-Methyl-4,6-dinitrophenol∣2-Methylnaphthalene∣2-Methylphenol∣2-Nitroaniline∣2-Nitrophenol∣3-Methylphenol (m-Cresol)∣3-Nitroaniline∣4-Chloro-3-methylphenol∣4-Chloroaniline∣4-Chlorophenyl phenyl ether∣4-Methylphenol (p-Cresol)∣4-Nitroaniline∣4-Nitrophenol∣Acenaphthene∣Acenaphthylene∣Adipic Acid Bis(2-ethylhexyl) Ester∣Aniline∣Anthracene∣Azobenzene∣Benz[a]anthracene∣Benzo(k)fluoranthene∣Benzo[a]pyrene∣Benzo[b]fluoranthene∣Benzo[ghi]perylene∣Benzyl alcohol∣Bis-(2-chloro-1-methylethyl)ether∣Bis(2-chloroethoxy)methane∣Bis-(2-chloroethyl)ether∣Carbazole∣Chrysene∣Dibenzo(a,h)anthracene∣Dibenzofuran∣Dibutyl phthalate∣Diethyl phthalate∣Di-n-octyl phthalate∣Diphenylamine∣Fluoranthene∣Fluorene∣Hexachlorobenzene∣Hexachlorobutadiene∣Hexachlorocyclopentadiene∣Hexachloroethane∣Indeno[1,2,3-cd]pyrene∣Isophorone∣Naphthalene∣Nitrobenzene∣N-Nitrosodimethylamine∣N-Nitroso-di-n-propylamine∣PBDE 3 (4-Bromodiphenyl Ether)∣Pentachlorophenol∣Phenanthrene∣Phenol∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-methyl ester∣Pyrene∣Pyridine |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dichloromethane |
| Quantity | 5 x 1 mL |
| Concentration or Composition (by Analyte or Components) | 1,2,4-Trichlorobenzene 1000 μg/mL∣1,2-Dichlorobenzene 1000 μg/mL∣1,2-Dinitrobenzene 1000 μg/mL∣1,3-Dichlorobenzene 1000 μg/mL∣1,3-Dinitrobenzene 1000 μg/mL∣1,4-Dichlorobenzene 1000 μg/mL∣1,4-Dinitrobenzene 1000 μg/mL∣1-Methylnaphthalene 1000 μg/mL∣2,3,4,6-Tetrachlorophenol 1000 μg/mL∣2,3,5,6-Tetrachlorophenol 1000 μg/mL∣2,4,5-Trichlorophenol 1000 μg/mL∣2,4,6-Trichlorophenol 1000 μg/mL∣2,4-Dichlorophenol 1000 μg/mL∣2,4-Dimethylphenol 1000 μg/mL∣2,4-Dinitrophenol 1000 μg/mL∣2,4-Dinitrotoluene 1000 μg/mL∣2,6-Dinitrotoluene 1000 μg/mL∣2-Chloronaphthalene 1000 μg/mL∣2-Chlorophenol 1000 μg/mL∣2-Methyl-4,6-dinitrophenol 1000 μg/mL∣2-Methylnaphthalene 1000 μg/mL∣2-Methylphenol 1000 μg/mL∣2-Nitroaniline 1000 μg/mL∣2-Nitrophenol 1000 μg/mL∣3-Methylphenol (m-Cresol) 1000 μg/mL∣3-Nitroaniline 1000 μg/mL∣4-Chloro-3-methylphenol 1000 μg/mL∣4-Chloroaniline 1000 μg/mL∣4-Chlorophenyl phenyl ether 1000 μg/mL∣4-Methylphenol (p-Cresol) 1000 μg/mL∣4-Nitroaniline 1000 μg/mL∣4-Nitrophenol 1000 μg/mL∣Acenaphthene 1000 μg/mL∣Acenaphthylene 1000 μg/mL∣Adipic Acid Bis(2-ethylhexyl) Ester 1000 μg/mL∣Aniline 1000 μg/mL∣Anthracene 1000 μg/mL∣Azobenzene 1000 μg/mL∣Benz[a]anthracene 1000 μg/mL∣Benzo(k)fluoranthene 1000 μg/mL∣Benzo[a]pyrene 1000 μg/mL∣Benzo[b]fluoranthene 1000 μg/mL∣Benzo[ghi]perylene 1000 μg/mL∣Benzyl alcohol 1000 μg/mL∣Bis-(2-chloro-1-methylethyl)ether 1000 μg/mL∣Bis(2-chloroethoxy)methane 1000 μg/mL∣Bis-(2-chloroethyl)ether 1000 μg/mL∣Carbazole 1000 μg/mL∣Chrysene 1000 μg/mL∣Dibenzo(a,h)anthracene 1000 μg/mL∣Dibenzofuran 1000 μg/mL∣Dibutyl phthalate 1000 μg/mL∣Diethyl phthalate 1000 μg/mL∣Di-n-octyl phthalate 1000 μg/mL∣Diphenylamine 1000 μg/mL∣Fluoranthene 1000 μg/mL∣Fluorene 1000 μg/mL∣Hexachlorobenzene 1000 μg/mL∣Hexachlorobutadiene 1000 μg/mL∣Hexachlorocyclopentadiene 1000 μg/mL∣Hexachloroethane 1000 μg/mL∣Indeno[1,2,3-cd]pyrene 1000 μg/mL∣Isophorone 1000 μg/mL∣Naphthalene 1000 μg/mL∣Nitrobenzene 1000 μg/mL∣N-Nitrosodimethylamine 1000 μg/mL∣N-Nitroso-di-n-propylamine 1000 μg/mL∣PBDE 3 (4-Bromodiphenyl Ether) 1000 μg/mL∣Pentachlorophenol 1000 μg/mL∣Phenanthrene 1000 μg/mL∣Phenol 1000 μg/mL∣Phthalic acid, benzylbutyl ester 1000 μg/mL∣Phthalic acid, bis-2-ethylhexyl ester 1000 μg/mL∣Phthalic acid, bis-methyl ester 1000 μg/mL∣Pyrene 1000 μg/mL∣Pyridine 1000 μg/mL |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto