missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 8270 Internal Standard Mixture 2000 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30315510
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30315510 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30315510 Supplier Dr. Ehrenstorfer Supplier No. DREGS09000428DI
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,4-Dichlorobenzene-D4∣1,4-Dioxane D8∣Acenaphthene D10∣Chrysene D12∣Naphthalene D8∣Perylene D12∣Phenanthrene D10
CAS 3855-82-1, 17647-74-4, 15067-26-2, 1719-03-5, 1146-65-2, 1520-96-3
Analyte or Component Names 1,4-Dichlorobenzene-D4∣1,4-Dioxane D8∣Acenaphthene D10∣Chrysene D12∣Naphthalene D8∣Perylene D12∣Phenanthrene D10
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dichloromethane
Quantity 5 x 1 mL
Concentration or Composition (by Analyte or Components) 1,4-Dioxane D8 1000 μg/mL∣1,4-Dichlorobenzene-D4 2000 μg/mL∣Acenaphthene D10 2000 μg/mL∣Chrysene D12 2000 μg/mL∣Naphthalene D8 2000 μg/mL∣Perylene D12 2000 μg/mL∣Phenanthrene D10 2000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.