missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 8270 LCS Mixture 100 μg/mL in Acetone:MeCl2 83.5:16.5, Dr. Ehrenstorfer

Product Code. 30321587
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30321587 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321587 Supplier Dr. Ehrenstorfer Supplier No. DREXA09000004AD

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2,4-Trichlorobenzene∣1,2-Dichlorobenzene∣1,2-Dinitrobenzene∣1,3-Dichlorobenzene∣1,3-Dinitrobenzene∣1,4-Dichlorobenzene∣1,4-Dinitrobenzene∣1-Methylnaphthalene∣2,3,4,6-Tetrachlorophenol∣2,3,5,6-Tetrachlorophenol∣2,4,5-Trichlorophenol∣2,4,6-Trichlorophenol∣2,4-Dichlorophenol∣2,4-Dimethylphenol∣2,4-Dinitrophenol∣2,4-Dinitrotoluene∣2,6-Dinitrotoluene∣2-Chloronaphthalene∣2-Chlorophenol∣2-Methyl-4,6-dinitrophenol∣2-Methylnaphthalene∣2-Methylphenol∣2-Nitroaniline∣2-Nitrophenol∣3,3'-Dichlorobenzidine∣3-Methylphenol (m-Cresol)∣3-Nitroaniline∣4-Chloro-3-methylphenol∣4-Chloroaniline∣4-Chlorophenyl phenyl ether∣4-Methylphenol (p-Cresol)∣4-Nitroaniline∣4-Nitrophenol∣Acenaphthene∣Acenaphthylene∣Adipic Acid Bis(2-ethylhexyl) Ester∣Aniline∣Anthracene∣Azobenzene∣Benz[a]anthracene∣Benzo(k)fluoranthene∣Benzo[a]pyrene∣Benzo[b]fluoranthene∣Benzo[ghi]perylene∣Benzoic acid∣Benzyl alcohol∣Bis-(2-chloro-1-methylethyl)ether∣Bis(2-chloroethoxy)methane∣Bis-(2-chloroethyl)ether∣Carbazole∣Chrysene∣Dibenzo(a,h)anthracene∣Dibenzofuran∣Dibutyl phthalate∣Diethyl phthalate∣Di-n-octyl phthalate∣Fluoranthene∣Fluorene∣Hexachlorobenzene∣Hexachlorobutadiene∣Hexachlorocyclopentadiene∣Hexachloroethane∣Indeno[1,2,3-cd]pyrene∣Isophorone∣Naphthalene∣Nitrobenzene∣N-Nitrosodimethylamine∣N-Nitroso-di-n-propylamine∣N-Nitroso-diphenylamine∣PBDE 3 (4-Bromodiphenyl Ether)∣Pentachlorophenol∣Phenanthrene∣Phenol∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-methyl ester∣Pyrene∣Pyridine
CAS 120-82-1, 95-50-1, 528-29-0, 541-73-1, 99-65-0, 106-46-7
Analyte or Component Names 1,2,4-Trichlorobenzene∣1,2-Dichlorobenzene∣1,2-Dinitrobenzene∣1,3-Dichlorobenzene∣1,3-Dinitrobenzene∣1,4-Dichlorobenzene∣1,4-Dinitrobenzene∣1-Methylnaphthalene∣2,3,4,6-Tetrachlorophenol∣2,3,5,6-Tetrachlorophenol∣2,4,5-Trichlorophenol∣2,4,6-Trichlorophenol∣2,4-Dichlorophenol∣2,4-Dimethylphenol∣2,4-Dinitrophenol∣2,4-Dinitrotoluene∣2,6-Dinitrotoluene∣2-Chloronaphthalene∣2-Chlorophenol∣2-Methyl-4,6-dinitrophenol∣2-Methylnaphthalene∣2-Methylphenol∣2-Nitroaniline∣2-Nitrophenol∣3,3'-Dichlorobenzidine∣3-Methylphenol (m-Cresol)∣3-Nitroaniline∣4-Chloro-3-methylphenol∣4-Chloroaniline∣4-Chlorophenyl phenyl ether∣4-Methylphenol (p-Cresol)∣4-Nitroaniline∣4-Nitrophenol∣Acenaphthene∣Acenaphthylene∣Adipic Acid Bis(2-ethylhexyl) Ester∣Aniline∣Anthracene∣Azobenzene∣Benz[a]anthracene∣Benzo(k)fluoranthene∣Benzo[a]pyrene∣Benzo[b]fluoranthene∣Benzo[ghi]perylene∣Benzoic acid∣Benzyl alcohol∣Bis-(2-chloro-1-methylethyl)ether∣Bis(2-chloroethoxy)methane∣Bis-(2-chloroethyl)ether∣Carbazole∣Chrysene∣Dibenzo(a,h)anthracene∣Dibenzofuran∣Dibutyl phthalate∣Diethyl phthalate∣Di-n-octyl phthalate∣Fluoranthene∣Fluorene∣Hexachlorobenzene∣Hexachlorobutadiene∣Hexachlorocyclopentadiene∣Hexachloroethane∣Indeno[1,2,3-cd]pyrene∣Isophorone∣Naphthalene∣Nitrobenzene∣N-Nitrosodimethylamine∣N-Nitroso-di-n-propylamine∣N-Nitroso-diphenylamine∣PBDE 3 (4-Bromodiphenyl Ether)∣Pentachlorophenol∣Phenanthrene∣Phenol∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-methyl ester∣Pyrene∣Pyridine
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetone/MeCl2
Industry Type Food Testing
Quantity 25 mL
Concentration or Composition (by Analyte or Components) 1,2,4-Trichlorobenzene 100 μg/mL∣1,2-Dichlorobenzene 100 μg/mL∣1,2-Dinitrobenzene 100 μg/mL∣1,3-Dichlorobenzene 100 μg/mL∣1,3-Dinitrobenzene 100 μg/mL∣1,4-Dichlorobenzene 100 μg/mL∣1,4-Dinitrobenzene 100 μg/mL∣1-Methylnaphthalene 100 μg/mL∣2,3,4,6-Tetrachlorophenol 100 μg/mL∣2,3,5,6-Tetrachlorophenol 100 μg/mL∣2,4,5-Trichlorophenol 100 μg/mL∣2,4,6-Trichlorophenol 100 μg/mL∣2,4-Dichlorophenol 100 μg/mL∣2,4-Dimethylphenol 100 μg/mL∣2,4-Dinitrophenol 100 μg/mL∣2,4-Dinitrotoluene 100 μg/mL∣2,6-Dinitrotoluene 100 μg/mL∣2-Chloronaphthalene 100 μg/mL∣2-Chlorophenol 100 μg/mL∣2-Methyl-4,6-dinitrophenol 100 μg/mL∣2-Methylnaphthalene 100 μg/mL∣2-Methylphenol 100 μg/mL∣2-Nitroaniline 100 μg/mL∣2-Nitrophenol 100 μg/mL∣3,3'-Dichlorobenzidine 100 μg/mL∣3-Methylphenol (m-Cresol) 100 μg/mL∣3-Nitroaniline 100 μg/mL∣4-Chloro-3-methylphenol 100 μg/mL∣4-Chloroaniline 100 μg/mL∣4-Chlorophenyl phenyl ether 100 μg/mL∣4-Methylphenol (p-Cresol) 100 μg/mL∣4-Nitroaniline 100 μg/mL∣4-Nitrophenol 100 μg/mL∣Acenaphthene 100 μg/mL∣Acenaphthylene 100 μg/mL∣Adipic Acid Bis(2-ethylhexyl) Ester 100 μg/mL∣Aniline 100 μg/mL∣Anthracene 100 μg/mL∣Azobenzene 100 μg/mL∣Benz[a]anthracene 100 μg/mL∣Benzo(k)fluoranthene 100 μg/mL∣Benzo[a]pyrene 100 μg/mL∣Benzo[b]fluoranthene 100 μg/mL∣Benzo[ghi]perylene 100 μg/mL∣Benzoic acid 100 μg/mL∣Benzyl alcohol 100 μg/mL∣Bis-(2-chloro-1-methylethyl)ether 100 μg/mL∣Bis(2-chloroethoxy)methane 100 μg/mL∣Bis-(2-chloroethyl)ether 100 μg/mL∣Carbazole 100 μg/mL∣Chrysene 100 μg/mL∣Dibenzo(a,h)anthracene 100 μg/mL∣Dibenzofuran 100 μg/mL∣Dibutyl phthalate 100 μg/mL∣Diethyl phthalate 100 μg/mL∣Di-n-octyl phthalate 100 μg/mL∣Fluoranthene 100 μg/mL∣Fluorene 100 μg/mL∣Hexachlorobenzene 100 μg/mL∣Hexachlorobutadiene 100 μg/mL∣Hexachlorocyclopentadiene 100 μg/mL∣Hexachloroethane 100 μg/mL∣Indeno[1,2,3-cd]pyrene 100 μg/mL∣Isophorone 100 μg/mL∣Naphthalene 100 μg/mL∣Nitrobenzene 100 μg/mL∣N-Nitrosodimethylamine 100 μg/mL∣N-Nitroso-di-n-propylamine 100 μg/mL∣N-Nitroso-diphenylamine 100 μg/mL∣PBDE 3 (4-Bromodiphenyl Ether) 100 μg/mL∣Pentachlorophenol 100 μg/mL∣Phenanthrene 100 μg/mL∣Phenol 100 μg/mL∣Phthalic acid, benzylbutyl ester 100 μg/mL∣Phthalic acid, bis-2-ethylhexyl ester 100 μg/mL∣Phthalic acid, bis-methyl ester 100 μg/mL∣Pyrene 100 μg/mL∣Pyridine 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.