missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epcam / CD326 Mouse, Clone: rVU-1D9, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 4072-MSM20-P1
Additional Details : Weight : 0.01000kg
Specifications
Epcam / CD326 | |
Cell surface | |
rVU-1D9 | |
Unconjugated | |
NP_002345 | |
Mouse | |
40-43kDa | |
Protein A/G Purified | |
RUO | |
4072 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
EPCAM | |
Small cell lung carcinoma cells | |
HT29 cells or breast tumor | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |