missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EpCAM/TROP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
226.00 € - 386.00 €
Specifications
| Antigen | EpCAM/TROP1 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 3 - 5 ug/mL, Immunohistochemistry-Paraffin 1:400 - 1:800 |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18151154
|
Novus Biologicals
NBP2-27107SS |
0.025 mg |
226.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18718733
|
Novus Biologicals
NBP2-27107 |
0.1 mg |
386.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EpCAM/TROP1 Polyclonal specifically detects EpCAM/TROP1 in Human, Mouse, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| EpCAM/TROP1 | |
| Western Blot 3 - 5 ug/mL, Immunohistochemistry-Paraffin 1:400 - 1:800 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Primate | |
| O55159 | |
| 4072 | |
| A portion of amino acids 250-350 of human EpCAM/CD326 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cancer Stem Cells, Tumor Biomarkers | |
| PBS with 0.05% Sodium Azide | |
| 17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2EGP34, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A | |
| EPCAM | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen for the antibody is 100% homologous in human, 94% in chimp and orangutan, 72% in pig, cow and mouse, 66% in dog and 61% homologous in rat. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title