Learn More
Invitrogen™ Epcoritamab Chimeric Recombinant Human Monoclonal Antibody
GREENER_CHOICE
Human Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA560133
 View more versions of this product
Product Code. 30283405
  • 582.00 € / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Epcoritamab (DuoBody™-CD3xCD20) is a proprietary bispecific antibody created using Genmab's DuoBody technology. It targets CD20 on the B-cells, a clinically well-validated target that is expressed in a wide variety of B-cell malignancies. Epcoritamab is currently under investigation in a Phase I/II study for multiple hematological B cell malignancies.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Epcoritamab Chimeric
Recombinant Monoclonal
Unconjugated
GEN3013
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized