Learn More
EphA2 Antibody (1C1), PE - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52677PE
Product Code. 18642008
  • 608.00 € / 0.10mL
Description
EphA2 Monoclonal antibody specifically detects EphA2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Agonist Activity, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EphA2
Monoclonal
PE
ARCC2, EC 2.7.10, EC 2.7.10.1, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK
1C1 was isolated after screening of a non-immunized human Fab phage library. Selection was based on the ability to bind soluble human recombinant EphA2 (residues 183-510). The Fab was subsequently converted to full length human IgG1.
0.1 mL
Cancer, Protein Kinase
1969
Store at 4°C in the dark.
IgG κ
Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Agonist Activity
1C1
PBS
Rabbit
Protein A purified
RUO
Primary
Human
Purified