Learn More
Ephrin-A1 Antibody (CL6501) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-76497PE
Product Code. 30297653
  • 737.00 € / 0.10mL
Description
Ephrin-A1 Monoclonal antibody specifically detects Ephrin-A1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ephrin-A1
Monoclonal
PE
ECKLG, EFL1, EPH-related receptor tyrosine kinase ligand 1, ephrin-A1, EPLG1TNF alpha-induced protein 4, Immediate early response protein B61, LERK1LERK-1, ligand of eph-related kinase 1, TNFAIP4B61, Tumor necrosis factor alpha-induced protein 4, tumor necrosis factor, alpha-induced protein 4
This antibody was developed using a recombinant protein derived from P20827, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer
1942
Store at 4⪚C in the dark.
IgG1
Western Blot
CL6501
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified