missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ephrin A3 Monoclonal Antibody (80814)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561326
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a member of the ephrin (EPH) family. The ephrins and EPH-related receptors comprise the largest subfamily of receptor protein-tyrosine kinases and have been implicated in mediating developmental events, especially in the nervous system and in erythropoiesis. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. This gene encodes an EFNA class ephrin.
Specifications
| Ephrin A3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P52797 | |
| EFNA3 | |
| Mouse myeloma cell line NS0-derived recombinant human Ephrin-A3, Asn31-Ser209. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 80814 | |
| Unconjugated | |
| EFNA3 | |
| AW494418; EFL2; EFL-2; EFNA3; EHK1 ligand; EHK1-L; EPH-related receptor tyrosine kinase ligand 3; ephrin A3; ephrin-A3; Epl3; EPLG3; Lerk3; LERK-3; ligand of eph-related kinase 3 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 1944 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction