missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ephrin A4 Monoclonal Antibody (80937)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561338
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Ephrin A4 is also known as LERK-4 or ELF-4. Ephrin-A4 binds EphA2, EphA3, EphA4, EphA5, EphA6, EphA7, and EphB1. To date, at least 14 members of the Eph receptor family and a family of 8 ligands have been identified. Ligands of Eph family receptors are structurally related membrane-bound proteins that can be subdivided into two major subclasses: ephrin-A and ephrin-B. Ligands in the ephrin-A subclass, including the prototype family member ephrin-A1 (B61), are membrane associated through glycosylphosphatidyl-inositollinkages; whereas ephrin-B subclass consists of ligands with transmembrane domains.
Specifications
| Ephrin A4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P52798 | |
| EFNA4 | |
| Mouse myeloma cell line NS0-derived recombinant human Ephrin-A4, Leu26-Gly171. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 80937 | |
| Unconjugated | |
| EFNA4 | |
| EFL4; EFL-4; Efna4; EPH-related receptor tyrosine kinase ligand 4; ephrin A4; ephrin-A4; Epl4; EPLG4; LERK4; LERK-4; ligand of eph-related kinase 4 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 1945 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction