missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Ephrin-A4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
199.00 € - 369.00 €
Specifications
| Antigen | Ephrin-A4 |
|---|---|
| Clone | 80949 |
| Dilution | Immunocytochemistry 8-25 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18743549
![]() |
R&D Systems
MAB3691-SP |
25 μg |
199.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18702758
![]() |
R&D Systems
MAB3691-100 |
100 μg |
369.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ephrin-A4 Monoclonal specifically detects Ephrin-A4 in Human samples. It is validated for Immunocytochemistry.Specifications
| Ephrin-A4 | |
| Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P52798 | |
| 1945 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 80949 | |
| Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| EFL4, EFL-4, EFNA4, EPH-related receptor tyrosine kinase ligand 4, ephrin-A4, EphrinA4, EPLG4MGC125826, LERK-4, LERK4FLJ57652, ligand of eph-related kinase 4 | |
| Mouse myeloma cell line NS0-derived recombinant human Ephrin-A4 Leu26-Gly171 Accession # P52798 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Ephrin-A4 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title