missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ephrin A4 Polyclonal Antibody

Product Code. 30534363
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30534363 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30534363 Supplier Invitrogen™ Supplier No. PA5148741

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

Ephrin A4 is also known as LERK-4 or ELF-4. Ephrin-A4 binds EphA2, EphA3, EphA4, EphA5, EphA6, EphA7, and EphB1. To date, at least 14 members of the Eph receptor family and a family of 8 ligands have been identified. Ligands of Eph family receptors are structurally related membrane-bound proteins that can be subdivided into two major subclasses: ephrin-A and ephrin-B. Ligands in the ephrin-A subclass, including the prototype family member ephrin-A1 (B61), are membrane associated through glycosylphosphatidyl-inositollinkages; whereas ephrin-B subclass consists of ligands with transmembrane domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ephrin A4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Gene EFNA4
Gene Accession No. P52798
Gene Alias EFL4; EFL-4; Efna4; EPH-related receptor tyrosine kinase ligand 4; ephrin A4; ephrin-A4; Epl4; EPLG4; LERK4; LERK-4; ligand of eph-related kinase 4
Gene Symbols EFNA4
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Ephrin-A4, Leu26-Gly171.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1945
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.