missing translation for 'onlineSavingsMsg'
Learn More

(-)-Epicatechin

Product Code. 15453839
Change view
Click to view available options
Quantity:
20 mg
Unit Size:
20mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15453839 20 mg 20mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15453839 Supplier Thermo Scientific Alfa Aesar Supplier No. J61218.F+
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 490-46-0 | C15H14O6 | 290.271 g/mol

An antioxidant and natural product from green tea

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 490-46-0
Melting Point 240°dec
Color White to Yellow
Molecular Formula C15H14O6
MDL Number MFCD00075648
Quantity 20 mg
Beilstein 92760
Synonym --epicatechin, epicatechin, l-epicatechin, --epicatechol, l-acacatechin, epicatechol, l-epicatechol, 2r,3r-2-3,4-dihydroxyphenyl chroman-3,5,7-triol, -epicatechin, 2r,3r---epicatechin
Solubility Information Soluble in water or alcohol
InChI Key PFTAWBLQPZVEMU-UKRRQHHQSA-N
SMILES C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C=C3)O)O)O
IUPAC Name (2R,3R)-2-(3,4-dihydroxyphenyl)-3,4-dihydro-2H-chromene-3,5,7-triol
Molecular Weight (g/mol) 290.271
PubChem CID 72276
ChEBI CHEBI:90
Formula Weight 290.27
Physical Form Crystalline Powder
Chemical Name or Material (-)-Epicatechin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.