missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ EpiLife™ CF Kit

Product Code. 11657124 Shop All Gibco Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11657124 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11657124 Supplier Gibco™ Supplier No. MEPICF500
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

EpiLife™ CF Kit

A sterile, liquid medium prepared without calcium chloride for the long-term, serum-free culture of human epidermal keratinocytes and human corneal epithelial cells. This basal medium requires the addition of calcium plus an appropriate supplement prior to use. Calcium chloride is provided as a separate component with each bottle of medium. EpiLife greatly extends the in vitro lifespan of human epidermal keratinocytes compared to other serum-free media systems. This basal media is animal origin free and contains essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts. It does not contain antibiotics, antimycotics, hormones, growth factors, or proteins. EpiLife™ medium is HEPES and bicarbonate buffered, and is designed for use in an incubator with an atmosphere of 5% CO2 and 95% air.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Keratinocyte Cells, Epithelial Cells
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Form Liquid
Product Type CF Kit
Serum Level Serum-free
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives HEPES, Phenol Red
Without Additives No Calcium Chloride
Product Line Cascade Biologics, EpiLife
Quantity 500 mL
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Culture Type Mammalian Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.