missing translation for 'onlineSavingsMsg'
Learn More

Epinephrine Sulfonic Acid, TRC

Product Code. 17982085
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17982085 100 mg 100mg
17971235 25 mg 25mg
17900835 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17982085 Supplier Toronto Research Chemicals Supplier No. TRCE588605100MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26405-77-6
Molecular Formula C9 H13 N O5 S
Molecular Weight (g/mol) 247.27
Synonym Benzenemethanesulfonic acid, 3,4-dihydroxy-α-[(methylamino)methyl]-, α-Toluenesulfonic acid, 3,4-dihydroxy-α-[(methylamino)methyl]- (6CI,8CI), 3,4-Dihydroxy-α-[(methylamino)methyl]benzenemethanesulfonic acid, 1-(3,4-Dihydroxy-phenyl)-2-methylamino-ethanesulfonic acid, 1-(3,4-Dihydroxyphenyl)-2-(methylamino)ethane-1-sulfonic acid, 1-(3,4-Dihydroxyphenyl)-2-(methylamino)ethanesulfonic acid, Epinephrine sulfonic acid, Adrenaline sulfonic acid
IUPAC Name 1-(3,4-dihydroxyphenyl)-2-(methylamino)ethanesulfonic acid
SMILES CNCC(c1ccc(O)c(O)c1)S(=O)(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H13NO5S/c1-10-5-9(16(13,14)15)6-2-3-7(11)8(12)4-6/h2-4,9-12H,5H2,1H3,(H,13,14,15)
Formula Weight 247.0514
Percent Purity >95%
Chemical Name or Material Epinephrine Sulfonic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.