missing translation for 'onlineSavingsMsg'
Learn More

Epinine HCl, MP Biomedicals

Product Code. 11409414 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11409414 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11409414 Supplier MP Biomedicals Supplier No. 0522265380
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 62-32-8
Molecular Formula C9H14ClNO2
MDL Number MFCD00035074
Quantity 100 mg
Synonym n-methyldopamine hydrochloride, methyldopamine hydrochloride, epinine hydrochloride, ephinine hydrochloride, deoxyepinephrine hydrochloride, unii-zv3mg8pax3, zv3mg8pax3, 3,4-dihydroxyphenylethylmethylamine hydrochloride, 4-2-methylaminoethyl pyrocatechol hydrochloride, 3,4-dihydroxyphenyl-1-methylamino-2-ethane hydrochloride
InChI Key JCDRZCWRRLKLTB-UHFFFAOYSA-N
SMILES [H+].[Cl-].CNCCC1=CC=C(O)C(O)=C1
IUPAC Name hydrogen 4-[2-(methylamino)ethyl]benzene-1,2-diol chloride
Molecular Weight (g/mol) 203.67
PubChem CID 86470
Formula Weight 203.67
Chemical Name or Material Epinine HCL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.