Learn More
Epirubicin hydrochloride, Thermo Scientific™
Cell-permeable anthracycline anti-tumor antibiotic
Brand:  Thermo Scientific Alfa Aesar J60411.MA
Quantity 10mg
 View more versions of this product
Product Code. 15403189
  • 248.00 € / 10mg
 Restricted Item
Estimated Shipment: 18-09-2026
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Identifiers
56390-09-1
579.983
MWWSFMDVAYGXBV-DPZLWMTJSA-N
123134690
CC1C(C(CC(O1)OC2CC(CC3=C(C4=C(C(=C23)O)C(=O)C5=C(C4=O)C=CC=C5OC)O)(C(=O)CO)O)N)O.Cl
C27H30ClNO11
MFCD00941448
Epiadriamycin
(7S,9S)-7-[(2R,4R,5R)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-9-(2-hydroxyacetyl)-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride
Specifications
56390-09-1
MFCD00941448
14,3623
Soluble in DMSO to 100mM, or in ethanol to 10mM
CC1C(C(CC(O1)OC2CC(CC3=C(C4=C(C(=C23)O)C(=O)C5=C(C4=O)C=CC=C5OC)O)(C(=O)CO)O)N)O.Cl
579.983
579.98
C27H30ClNO11
10mg
Epiadriamycin
MWWSFMDVAYGXBV-DPZLWMTJSA-N
(7S,9S)-7-[(2R,4R,5R)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-9-(2-hydroxyacetyl)-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride
123134690
Epirubicin hydrochloride
Product Suggestions
Product Suggestions
Videos
SDS Documents