accessibility menu, dialog, popup

UserName

ClH3COOOH(S)OH(S)O(R)(S)NH2(R)OH(S)CH3OOHOOOHH

CAS RN 56390-09-1

IUPAC Name: hydrogen (8S,10S)-10-{[(2R,4S,5R,6S)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy}-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-5,7,8,9,10,12-hexahydrotetracene-5,12-dione chloride
Synonyms: hydrogen epirubicin chloride
Molecular Weight (g/mol): 579.98
Molecular Formula: C27H30ClNO11
InChi Key: MWWSFMDVAYGXBV-FGBSZODSSA-N
SMILES: [H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3O[C@H]3C[C@H](N)[C@@H](O)[C@H](C)O3)C(=O)CO)C(O)=C1C2=O
Molecular Weight (g/mol) 
  • (10)
  • (2)
Quantity 
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (3)
  • (4)
  • (10)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (6)
  • (4)
  • (4)
  • (2)
  • (1)
  • (1)