missing translation for 'onlineSavingsMsg'
Learn More

Epirubicin hydrochloride

Product Code. 15413189
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15413189 100 mg 100mg
15403189 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15413189 Supplier Thermo Scientific Alfa Aesar Supplier No. J60411.MC
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cell-permeable anthracycline anti-tumor antibiotic | CAS: 56390-09-1 | C27H30ClNO11 | 579.983 g/mol

Cell-permeable anthracycline anti-tumor antibiotic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56390-09-1
Molecular Formula C27H30ClNO11
Molecular Weight (g/mol) 579.983
MDL Number MFCD00941448
InChI Key MWWSFMDVAYGXBV-DPZLWMTJSA-N
Synonym Epiadriamycin
PubChem CID 123134690
IUPAC Name (7S,9S)-7-[(2R,4R,5R)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-9-(2-hydroxyacetyl)-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride
SMILES CC1C(C(CC(O1)OC2CC(CC3=C(C4=C(C(=C23)O)C(=O)C5=C(C4=O)C=CC=C5OC)O)(C(=O)CO)O)N)O.Cl

Specifications

Melting Point 185°C
Color Red
Quantity 100 mg
Merck Index 14,3623
Solubility Information Soluble in DMSO to 100mM, or in ethanol to 10mM
Formula Weight 579.98
Physical Form Solid
Chemical Name or Material Epirubicin hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.