missing translation for 'onlineSavingsMsg'
Learn More
Learn More
epithelial Sodium Channel beta Antibody (14E10), Novus Biologicals™
Mouse Monoclonal Antibody
166.00 € - 542.00 €
Specifications
| Antigen | epithelial Sodium Channel beta |
|---|---|
| Clone | 140000000000 |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:150 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18242562
|
Novus Biologicals
NBP2-59383 |
100 μg |
542.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18665406
|
Novus Biologicals
NBP2-59383-25ug |
25 μg |
166.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
epithelial Sodium Channel beta Monoclonal specifically detects epithelial Sodium Channel beta in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| epithelial Sodium Channel beta | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:150 | |
| Monoclonal | |
| Purified | |
| RUO | |
| amiloride-sensitive sodium channel subunit beta, amiloride-sensitive sodium channel subunit beta 1, BESC1, Beta-ENaC, beta-NaCH, ENaCB, ENaCbeta, Epithelial Na(+) channel subunit beta, epithelial sodium channel beta-2 subunit, epithelial sodium channel beta-3 subunit, nasal epithelial sodium channel beta subunit, Nonvoltage-gated sodium channel 1 subunit beta, SCNEB, sodium channel, nonvoltage-gated 1, beta | |
| SCNN1B | |
| IgG1 | |
| Protein G purified |
| 140000000000 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Phospho Specific | |
| 6338 | |
| Synthetic peptide from the N-terminal of Rat ENaC alpha (aa. 46-68) | |
| Primary | |
| Detects 85kDa. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title