missing translation for 'onlineSavingsMsg'
Learn More

Epothilone B, MedChemExpress

Product Code. 16493287
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493287 50 mg 50mg
16437278 10 mg 10mg
16430296 2 mg 2mg
16419797 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16493287 Supplier Medchem Express Supplier No. HY17029
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Epothilone B is a microtubule stabilizer with a Ki of 0.71μM. It acts by binding to the αβ-tubulin heterodimer subunit which causes decreasing of αβ-tubulin dissociation.

Epothilone B is a microtubule stabilizer with a Ki of 0.71μM. It acts by binding to the αβ-tubulin heterodimer subunit which causes decreasing of αβ-tubulin dissociation.

Specifications

Chemical Name or Material Epothilone B
Physical Form Powder
CAS 152044-54-7
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C27H41NO6S
For Use With (Application) Cancer-programmed cell death
Synonym EPO 906 Patupilone
Solubility Information DMSO : ≥ 125 mg/mL (246.22 mM)
SMILES O=C(C[C@H](O)C1(C)C)O[C@H](/C(C)=C/C2=CSC(C)=N2)C[C@]3([H])O[C@]3(C)CCC[C@H](C)[C@H](O)[C@@H](C)C1=O
Molecular Weight (g/mol) 507.68
Formula Weight 507.68
Percent Purity 98.97%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.