missing translation for 'onlineSavingsMsg'
Learn More

Epothilone D, MedChemExpress

Product Code. 16466418
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16466418 10 mg 10mg
16463466 25 mg 25mg
16449007 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16466418 Supplier Medchem Express Supplier No. HY15278
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Epothilone D (KOS 862) is a potent microtubule stabilizer.

Epothilone D (KOS 862) is a potent microtubule stabilizer.

Specifications

Chemical Name or Material Epothilone D
Physical Form Powder
CAS 189453-10-9
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C27H41NO5S
For Use With (Application) Cancer-programmed cell death
Synonym KOS 862
Solubility Information DMSO : ≥ 100 mg/mL (203.38 mM)
SMILES O=C(O[C@@H](C/C=C(CCC[C@@H]([C@@H]([C@H](C1=O)C)O)C)/C)/C(C)=C/C2=CSC(C)=N2)C[C@@H](C1(C)C)O
Molecular Weight (g/mol) 491.68
Formula Weight 491.68
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.