Learn More
Eppendorf™ Storage Foil Adhesive Seal for Microplates
Product Code. 11803482 Shop All Eppendorf Products
Change view
Click to view available options
Material:
Aluminum
Unit Size:
Pack of 100
Product Code. Material Unit Size
11803482 Aluminum Pack of 100
1 options
Product Code. 11803482 Supplier Eppendorf™ Supplier No. 0030127889
  • ONLINE EXCLUSIVE PRICE
    153.00 €
    143.00 € / Pack of 100
    Save 10.00 €
    6% Off
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
Bulk Quote Request
missing translation for 'validMainframeMsg'
Storage Film and Foil glued seals for simple and dependable sealing during sample storage
  • Heat Sealing Film and Foil for continuous locking with the best evaporation protection
  • Foil products are made of aluminum, can be pierced, and protect light-sensitive samples
  • Film products are transparent and provide protection against unwanted punctures
  • Eppendorf Plate Lid: stable, flexible protection of samples during short-term storage
TRUSTED_SUSTAINABILITY
Specifications
Material Aluminum
Product Type Storage Foil