missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ twin.tec™ PCR Plates 384-well, 45 μL - PCR Plates
GREENER_CHOICE

Product Code. p-2713152 Shop All Eppendorf Products
Click to view available options
Color:
Blue
Colorless
Barcode:
No Barcode
Packaging:
Individually Blister Wrapped
Purity or Quality Grade:
Forensic DNA Grade (Non-sterile)
PCR Clean (Non-sterile)
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 10023927

Brand: Eppendorf™ 0030129342

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Eppendorf's twin.tec™ PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. With raised well rims, these plates enable an effective seal, minimizing the risk of cross-contamination and contributing to consistently reliable results.

  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • Improved well-to-well tolerance.
  • Cut-away corner and alphanumeric grid-referencing.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.
  • Batch-tested and certified free of DNA, DNase, RNase and PCR inhibitors (PCR clean).
  • Safe Code option provides 3-level coding for safe sample and vessel identification.
  • ECC 200 error correction function enables safe code reading with up to 30% damaged codes.

Certifications

Certified free of human DNA, DNase, RNase, and PCR inhibitors

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance Free of Human DNA, DNase, RNase and PCR Inhibitors
Format Standard
Array 16 x 24
Autoclavable Autoclavable
Packaging Individually Blister Wrapped
Color Colorless
Product Type PCR Plate
For Use With (Equipment) Automated Systems
Skirt Style skirted
Material Polypropylene, Polycarbonate
Volume (Metric) Well 45 μL
No. of Wells 384
Sterility Non-sterile
Well Shape Round Bottom (U-well)
Max. RCF 2250 x g
Purity or Quality Grade PCR Clean (Non-sterile)
Quantity 10 Pk.
Well Color Colorless
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.