missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals EPPS, 0.2M buffer soln., pH 8.0

Product Code. 15463909
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15463909 100 mL 100mL
15473909 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15463909 Supplier Thermo Scientific Chemicals Supplier No. J61296.AE
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 16052-06-5 | C9H20N2O4S | 252.329 g/mol

EPPS or HEPPS is a commonly used Good’s buffer. It is often used as separator in ultrathin isoelectric focusing gels and enhances the resolution of phosphoglucomutase. It is used as a buffering agent in biology and biochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
EPPS or HEPPS is a commonly used Good’s buffer. It is often used as separator in ultrathin isoelectric focusing gels and enhances the resolution of phosphoglucomutase. It is used as a buffering agent in biology and biochemistry.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C9H20N2O4S
Molecular Weight (g/mol) 252.329
MDL Number MFCD00006160
InChI Key OWXMKDGYPWMGEB-UHFFFAOYSA-N
Synonym HEPPS
PubChem CID 85255
ChEBI CHEBI:42298
IUPAC Name 3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
SMILES C1CN(CCN1CCCS(=O)(=O)O)CCO

Specifications

Concentration 0.2 M
pH 8.0
Physical Form Liquid
Quantity 100 mL
Solubility Information It is soluble in water.
Chemical Name or Material EPPS buffer soln.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.