missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals EPPS, 99%, for biochemistry

Product Code. 10104480 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10104480 100 g Glass bottle 100g
10474205 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10104480 Supplier Thermo Scientific Chemicals Supplier No. 172601000
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16052-06-5 | C9H20N2O4S | 252.329 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16052-06-5
Molecular Formula C9H20N2O4S
Molecular Weight (g/mol) 252.329
MDL Number MFCD00006160
InChI Key OWXMKDGYPWMGEB-UHFFFAOYSA-N
Synonym 3-[4-(2-Hydroxyethyl)-1-piperazine]propanesulfonic acid
PubChem CID 85255
ChEBI CHEBI:42298
IUPAC Name 3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
SMILES C1CN(CCN1CCCS(=O)(=O)O)CCO

Specifications

Absorbance (0.1 M aq. soln.) (1 cm cell)
Absorbance Conditions (0.1M aq. soln.) (1 cm cell)
Color White
pH 5 to 6 (10% w/v soln. at 20°C)
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Formula Weight 252.33
Percent Purity ≥98.5%
Grade Biochemical
Chemical Name or Material EPPS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.