missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Epstein Barr Virus LMP1 Monoclonal Antibody (EBS-I-025)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA533322
This item is not returnable.
View return policy
Description
EBS-I-025 reacts with a conserved repeat domain on LMP1 (AA 268-286), present in all EBV isolates. LMP1 is expressed in most viral latency stages in vitro and in vivo.
Epstein-Barr virus (EBV) is a human herpes virus, which is associated with conditions such as Hodgkin's disease and Burkitt's Lymphoma and is the causative agent in mononucleosis in adolescents. EBV latently infects B lymphocytes. Infected B cells express EBV nuclear antigens and latent proteins LMP1, LMP2A and LMP2B. LMP2A forms aggregates in the plasma membranes of B lymphocytes, where it functions as a negative regulator of the Src and Syk protein tyrosine kinases. Studies show that LMP2A blocks B-cell receptor (BCR) signal transduction in EBV immortalized B cells in vitro and may play an important role in maintaining a latent EBV infection within the peripheral blood B cells of infected individuals.EBNA-1 is the only viral protein consistently expressed in all malignancies associated with EBV.
Specifications
| Epstein Barr Virus LMP1 | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.02% sodium azide | |
| Mouse | |
| Purified | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| EBS-I-025 | |
| Unconjugated | |
| EBV | |
| LMP1 b-galactosidase fusion protein (AA 198-486). | |
| 100 μg | |
| Primary | |
| 4°C, do not freeze | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction