missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ER alpha/NR3A1 Antibody (ESR1/7444R), Novus Biologicals™
Rabbit Monoclonal Antibody
286.00 € - 530.00 €
Specifications
| Antigen | ER alpha/NR3A1 |
|---|---|
| Clone | ESR1/7444R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18664445
|
Novus Biologicals
NBP3-20628-100ug |
100 μg |
530.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18698464
|
Novus Biologicals
NBP3-20628-20ug |
20 μg |
286.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ER alpha/NR3A1 Monoclonal antibody specifically detects ER alpha/NR3A1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| ER alpha/NR3A1 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 10mM PBS with 0.05% BSA | |
| 2099 | |
| IgG κ | |
| Protein A or G purified |
| ESR1/7444R | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ER, ER alpha, Era, ER-alpha, ESRESRA, Estradiol receptor, estrogen receptor, estrogen receptor 1, estrogen receptor alpha, estrogen receptor alpha delta 3*4,56,7*/819-2 isoform, estrogen receptor alpha delta 4 +49 isoform, estrogen receptor alpha delta 4*5,6,7*/654 isoform, NR3A1DKFZp686N23123, Nuclear receptor subfamily 3 group A member 1 | |
| Recombinant fragment (around aa495-595) of human ER alpha/NR3A1 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title