Learn More
ER alpha/NR3A1, p Ser106 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-81912-0.025ML
 View more versions of this product
Product Code. 18280372
  • 270.00 € / 25µL
Description
ER alpha/NR3A1 Polyclonal specifically detects ER alpha/NR3A1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ER alpha/NR3A1 (p Ser106)
Polyclonal
Unconjugated
phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide
ER, ER alpha, Era, ER-alpha, ESRESRA, Estradiol receptor, estrogen receptor, estrogen receptor 1, estrogen receptor alpha, estrogen receptor alpha delta 3*4,56,7*/819-2 isoform, estrogen receptor alpha delta 4 +49 isoform, estrogen receptor alpha delta 4*5,6,7*/654 isoform, NR3A1DKFZp686N23123, Nuclear receptor subfamily 3 group A member 1
Rabbit
66 kDa
0.025 mL
Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
2099
Rat, Human, Mouse
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence
1.0 mg/mL
Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100
P03372
ESR1
The antiserum was produced against synthesized phosphopeptide derived from human Estrogen Receptor- alpha around the phosphorylation site of serine 106 (S-P- SP-P-L).
Affinity Purified
RUO
Primary
Estrogen Receptor alpha (specific to Phospho-Ser106) detects endogenous levels of Estrogen Receptor only when phosphorylated at serine 106.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.