missing translation for 'onlineSavingsMsg'
Learn More

ERAB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16837163 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16837163 20 μL 20µL
16827163 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16837163 Supplier Proteintech Supplier No. 106481AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes 3-hydroxyacyl-CoA dehydrogenase type II, a member of the short-chain dehydrogenase/reductase superfamily. The gene product is a mitochondrial protein that catalyzes the oxidation of a wide variety of fatty acids, alcohols, and steroids. The protein has been implicated in the development of Alzheimer′s disease, and mutations in the gene are the cause of 2-methyl-3-hydroxybutyryl-CoA dehydrogenase deficiency (MHBD). Several alternatively spliced transcript variants have been identified, but the full-length nature of only two transcript variants has been determined.

Specifications

Antigen ERAB
Applications Immunoprecipitation, Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HSD17B10
Gene Accession No. O08756, O70351, Q99714
Gene Alias 17 beta HSD 10, 17b HSD10, ABAD, CAMR, DUPXp11.22, ERAB, HADH2, HCD2, HSD17B10, MHBD, MRPP2, MRX17, MRX31, MRXS10, SCHAD, SDR5C1, Type II HADH, XH98G2
Gene Symbols Hsd17b10
Host Species Rabbit
Immunogen ERAB Fusion Protein Ag1020
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15108, 3028, 63864
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.