missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB2/Her2 Antibody (HRB2/273), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
552.00 € - 595.00 €
Specifications
| Antigen | ErbB2/Her2 |
|---|---|
| Clone | HRB2/273 |
| Applications | Flow Cytometry, ELISA, Immunofluorescence, Immunofluorescence, CyTOF |
| Classification | Monoclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18678638
|
Novus Biologicals
NBP2-48005B-100ul |
100 μg |
595.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18698927
|
Novus Biologicals
NBP2-48005B |
0.1 mL |
552.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ErbB2/Her2 Monoclonal antibody specifically detects ErbB2/Her2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, CyTOFSpecifications
| ErbB2/Her2 | |
| Flow Cytometry, ELISA, Immunofluorescence, Immunofluorescence, CyTOF | |
| Biotin | |
| Mouse | |
| Breast Cancer, Cancer, Cellular Markers, Core ESC Like Genes, Oncogenes, Protein Kinase, Stem Cell Markers, Tumor Suppressors | |
| PBS | |
| 2064 | |
| IgG1 κ | |
| Protein A or G purified |
| HRB2/273 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD340, CD340 antigen, c-erb B2/neu protein, EC 2.7.10, EGFR2, HER-2, HER2EC 2.7.10.1, herstatin, Metastatic lymph node gene 19 protein, MLN 19, MLN19, NEUHER-2/neu, neuroblastoma/glioblastoma derived oncogene homolog, NGLTKR1, p185erbB2, Proto-oncogene c-ErbB-2, Proto-oncogene Neu, receptor tyrosine-protein kinase erbB-2, Tyrosine kinase-type cell surface receptor HER2, v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2(neuro/glioblastoma derived oncogene homolog), v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastomaderived oncogene homolog (avian) | |
| Recombinant extracellular domain of human ErbB2/Her2 protein (Uniprot: P04626) | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title