missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERCC8 Monoclonal Antibody (OTI5C9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808545
This item is not returnable.
View return policy
Description
This gene encodes a WD repeat protein, which interacts with Cockayne syndrome type B (CSB) protein and with p44 protein, a subunit of the RNA polymerase II transcription factor IIH. Mutations in this gene have been identified in patients with hereditary disease Cockayne syndrome (CS). CS cells are abnormally sensitive to ultraviolet radiation and are defective in the repair of transcriptionally active genes.Specifications
| ERCC8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13216 | |
| ERCC8 | |
| Full length human recombit protein of human ERCC8 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5C9 | |
| Unconjugated | |
| ERCC8 | |
| CKN1, CSA, UVSS2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1161 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction