missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERK1 Antibody (1E5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22203-0.025ml
This item is not returnable.
View return policy
Description
ERK1 Monoclonal specifically detects ERK1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ERK1 | |
| Monoclonal | |
| Unconjugated | |
| EC 2.7.11, ERK-1, ERK1p44-MAPK, ERT2, Extracellular signal-regulated kinase 1, extracellular signal-related kinase 1, HS44KDAP, HUMKER1A, Insulin-stimulated MAP2 kinase, MAP kinase 1, MAP kinase 3, MAP kinase isoform p44, MAPK 1, MAPK 3, MGC20180, Microtubule-associated protein 2 kinase, Mitogen-activated protein kinase 1, mitogen-activated protein kinase 3, p44erk1, p44-ERK1, p44mapk, PRKM3EC 2.7.11.24 | |
| Mouse | |
| 44 kDa | |
| 0.025 mL | |
| Angiogenesis, Breast Cancer, Cancer, Cell Cycle and Replication, Diabetes Research, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Signal Transduction | |
| 5595 | |
| Human, Mouse, Rat, Monkey | |
| Ascites |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 100000 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| MAPK3 | |
| Purified recombinant fragment of human ERK1 expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| The monoclonal antibody 6A7 reacts with human, mouse, monkey and rat Bax but does not bind the soluble cytosolic form of Bax; however, treatment of cells with non-ionic detergents exposes the epitope and allows binding of 6A7 to monomeric forms of Bax but not Bax complexed with either Bcl-2 or Bcl-xL. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction