missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ERK1 Inhibitor Peptide Set

Product Code. 18110564 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
2 mg
5 mg
Unit Size:
2mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18110564 5 mg 5mg
18116222 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18110564 Supplier Novus Biologicals™ Supplier No. NBP2293335MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use in research applications

TRUSTED_SUSTAINABILITY

Specifications

Host Species Human, Mouse, Rat, Hamster, Rabbit, Xenopus
Components ERK Inhibitor peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKKGMPKKKPTPIQLN (ERK inhibitor sequence: GMPKKKPTPIQLN). Molecular weight: 3795. Antennapedia Control peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361
For Use With (Application) Inhibition of Erk activation
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Quantity 5 mg
Product Type ERK1 Inhibitor Peptide Set
Molecular Weight (g/mol) 3795
Inhibitors ERK1
Form Lyophilized

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.